CAS RN 7558-79-4
Sodium Phosphate Dibasic Anhydrous (Granular or Powder/Certified ACS), Fisher Chemical™
CAS: 7558-79-4 Molecular Formula: HNa2O4P Molecular Weight (g/mol): 141.96 MDL Number: MFCD00003496 InChI Key: BNIILDVGGAEEIG-UHFFFAOYSA-L Synonym: disodium hydrogen phosphate,disodium phosphate,disodium hydrogenorthophosphate,sodium phosphate dibasic,dibasic sodium phosphate,acetest,soda phosphate,disodium orthophosphate,sodium hydrogen phosphate,phosphoric acid, disodium salt PubChem CID: 24203 ChEBI: CHEBI:34683 IUPAC Name: disodium hydrogen phosphate SMILES: [Na+].[Na+].OP([O-])([O-])=O
Sodium Phosphate Dibasic Anhydrous (White Granules or Powder), Fisher BioReagents™
CAS: 7558-79-4 Molecular Formula: HNa2O4P Molecular Weight (g/mol): 141.96 MDL Number: MFCD00003496 InChI Key: BNIILDVGGAEEIG-UHFFFAOYSA-L Synonym: disodium hydrogen phosphate,disodium phosphate,disodium hydrogenorthophosphate,sodium phosphate dibasic,dibasic sodium phosphate,acetest,soda phosphate,disodium orthophosphate,sodium hydrogen phosphate,phosphoric acid, disodium salt PubChem CID: 24203 ChEBI: CHEBI:34683 SMILES: [Na+].[Na+].OP([O-])([O-])=O
Sodium Phosphate Dibasic Anhydrous (USP), Fisher Chemical™
CAS: 7558-79-4 Molecular Formula: HNa2O4P Molecular Weight (g/mol): 141.96 MDL Number: MFCD00003496 InChI Key: BNIILDVGGAEEIG-UHFFFAOYSA-L Synonym: disodium hydrogen phosphate,disodium phosphate,disodium hydrogenorthophosphate,sodium phosphate dibasic,dibasic sodium phosphate,acetest,soda phosphate,disodium orthophosphate,sodium hydrogen phosphate,phosphoric acid, disodium salt PubChem CID: 24203 ChEBI: CHEBI:34683 IUPAC Name: disodium hydrogen phosphate SMILES: [Na+].[Na+].OP([O-])([O-])=O
Sodium Phosphate Dibasic Anhydrous (Powder/Laboratory), Fisher Chemical
CAS: 7558-79-4 Molecular Formula: HNa2O4P Molecular Weight (g/mol): 141.96 MDL Number: MFCD00003496 InChI Key: BNIILDVGGAEEIG-UHFFFAOYSA-L Synonym: disodium hydrogen phosphate,disodium phosphate,disodium hydrogenorthophosphate,sodium phosphate dibasic,dibasic sodium phosphate,acetest,soda phosphate,disodium orthophosphate,sodium hydrogen phosphate,phosphoric acid, disodium salt PubChem CID: 24203 ChEBI: CHEBI:34683 IUPAC Name: disodium hydrogen phosphate SMILES: [Na+].[Na+].OP([O-])([O-])=O
Sodium Phosphate Dibasic Anhydrous (USP/FCC/EP), Fisher Chemical™
CAS: 7558-79-4 Molecular Formula: HNa2O4P Molecular Weight (g/mol): 141.96 MDL Number: MFCD00003496 InChI Key: BNIILDVGGAEEIG-UHFFFAOYSA-L Synonym: disodium hydrogen phosphate,disodium phosphate,disodium hydrogenorthophosphate,sodium phosphate dibasic,dibasic sodium phosphate,acetest,soda phosphate,disodium orthophosphate,sodium hydrogen phosphate,phosphoric acid, disodium salt PubChem CID: 24203 ChEBI: CHEBI:34683 IUPAC Name: disodium hydrogen phosphate SMILES: [Na+].[Na+].OP([O-])([O-])=O
Sodium hydrogen phosphate, anhydrous, ACS, 99.0% min
CAS: 7558-79-4 Molecular Formula: HNa2O4P Molecular Weight (g/mol): 141.96 MDL Number: MFCD00003496 InChI Key: BNIILDVGGAEEIG-UHFFFAOYSA-L Synonym: disodium hydrogen phosphate,disodium phosphate,disodium hydrogenorthophosphate,sodium phosphate dibasic,dibasic sodium phosphate,acetest,soda phosphate,disodium orthophosphate,sodium hydrogen phosphate,phosphoric acid, disodium salt PubChem CID: 24203 ChEBI: CHEBI:34683 IUPAC Name: disodium;hydrogen phosphate SMILES: [Na+].[Na+].OP([O-])([O-])=O
MilliporeSigma™ Sodium Phosphate, Dibasic, ACS Reagent Grade, Calbiochem™,
CAS: 7558-79-4 Molecular Formula: HNa2O4P Molecular Weight (g/mol): 141.96 MDL Number: MFCD00003496 InChI Key: BNIILDVGGAEEIG-UHFFFAOYSA-L Synonym: Na2HPO4 PubChem CID: 24203 ChEBI: CHEBI:34683 IUPAC Name: disodium hydrogen phosphate SMILES: [Na+].[Na+].OP([O-])([O-])=O
Sodium hydrogen phosphate, 98+%
CAS: 7558-79-4 Molecular Formula: HNa2O4P Molecular Weight (g/mol): 141.96 MDL Number: MFCD00003496 InChI Key: BNIILDVGGAEEIG-UHFFFAOYSA-L Synonym: disodium hydrogen phosphate,disodium phosphate,disodium hydrogenorthophosphate,sodium phosphate dibasic,dibasic sodium phosphate,acetest,soda phosphate,disodium orthophosphate,sodium hydrogen phosphate,phosphoric acid, disodium salt PubChem CID: 24203 ChEBI: CHEBI:34683 SMILES: [Na+].[Na+].OP([O-])([O-])=O
Buffer solution pH 7, Phosphate buffer
CAS: 7558-79-4 | H2KO4P