CAS RN 111-30-8
Glutaraldehyde Solution, 25% (Certified), Fisher Chemical™
CAS: 111-30-8 Molecular Formula: C5H8O2 Molecular Weight (g/mol): 100.12 MDL Number: MFCD00007025 InChI Key: SXRSQZLOMIGNAQ-UHFFFAOYSA-N Synonym: glutaraldehyde,glutaral,glutaric dialdehyde,cidex,glutardialdehyde,1,5-pentanedial,sonacide,glutarol,aldesan,glutaric aldehyde PubChem CID: 3485 ChEBI: CHEBI:64276 IUPAC Name: pentanedial SMILES: O=CCCCC=O
Glutaric dialdehyde, 8% solution in water
CAS: 111-30-8 | C5H8O2 | 100.12 g/mol
Glutaric dialdehyde, 50 wt.% solution in water
CAS: 111-30-8 | C5H8O2 | 100.12 g/mol
Glutaric dialdehyde, electron microscopy grade, 25% solution in water, purified
CAS: 111-30-8 | C5H8O2 | 100.12 g/mol
Glutaraldehyde, 25% aq. soln.
CAS: 111-30-8 Molecular Formula: C5H8O2 Molecular Weight (g/mol): 100.12 MDL Number: MFCD00007025 InChI Key: SXRSQZLOMIGNAQ-UHFFFAOYSA-N Synonym: glutaraldehyde,glutaral,glutaric dialdehyde,cidex,glutardialdehyde,1,5-pentanedial,sonacide,glutarol,aldesan,glutaric aldehyde PubChem CID: 3485 ChEBI: CHEBI:64276 IUPAC Name: pentanedial SMILES: O=CCCCC=O
Glutaric dialdehyde, 25% w/v solution in water
CAS: 111-30-8 | C5H8O2 | 100.12 g/mol
Glutaraldehyde, 50% aq. soln.
CAS: 111-30-8 Molecular Formula: C5H8O2 Molecular Weight (g/mol): 100.12 MDL Number: MFCD00007025 InChI Key: SXRSQZLOMIGNAQ-UHFFFAOYSA-N Synonym: glutaraldehyde,glutaral,glutaric dialdehyde,cidex,glutardialdehyde,1,5-pentanedial,sonacide,glutarol,aldesan,glutaric aldehyde PubChem CID: 3485 ChEBI: CHEBI:64276 IUPAC Name: pentanedial SMILES: O=CCCCC=O
Glutaraldehyde (24-26% in Water), TCI America™
CAS: 111-30-8 Molecular Formula: C5H8O2 Molecular Weight (g/mol): 100.12 MDL Number: MFCD00007025 InChI Key: SXRSQZLOMIGNAQ-UHFFFAOYSA-N Synonym: glutaraldehyde,glutaral,glutaric dialdehyde,cidex,glutardialdehyde,1,5-pentanedial,sonacide,glutarol,aldesan,glutaric aldehyde PubChem CID: 3485 ChEBI: CHEBI:64276 IUPAC Name: pentanedial SMILES: O=CCCCC=O
Glutaraldehyde, 50% Aqueous Solution, MP Biomedicals
CAS: 111-30-8 Molecular Formula: C5H8O2 Molecular Weight (g/mol): 100.12 MDL Number: MFCD00007025 InChI Key: SXRSQZLOMIGNAQ-UHFFFAOYSA-N Synonym: glutaraldehyde,glutaral,glutaric dialdehyde,cidex,glutardialdehyde,1,5-pentanedial,sonacide,glutarol,aldesan,glutaric aldehyde PubChem CID: 3485 ChEBI: CHEBI:64276 IUPAC Name: pentanedial SMILES: O=CCCCC=O
Glutaraldehyde, 25% aqueous solution, MP Biomedicals™
CAS: 111-30-8 Molecular Formula: C5H8O2 Molecular Weight (g/mol): 100.12 MDL Number: MFCD00007025 InChI Key: SXRSQZLOMIGNAQ-UHFFFAOYSA-N Synonym: glutaraldehyde,glutaral,glutaric dialdehyde,cidex,glutardialdehyde,1,5-pentanedial,sonacide,glutarol,aldesan,glutaric aldehyde PubChem CID: 3485 ChEBI: CHEBI:64276 IUPAC Name: pentanedial SMILES: O=CCCCC=O
Glutaraldehyde (ca. 50% in Water, ca. 5.6mol/L), TCI America™
CAS: 111-30-8 Molecular Formula: C5H8O2 Molecular Weight (g/mol): 100.12 MDL Number: MFCD00007025 InChI Key: SXRSQZLOMIGNAQ-UHFFFAOYSA-N Synonym: glutaraldehyde,glutaral,glutaric dialdehyde,cidex,glutardialdehyde,1,5-pentanedial,sonacide,glutarol,aldesan,glutaric aldehyde PubChem CID: 3485 ChEBI: CHEBI:64276 IUPAC Name: pentanedial SMILES: O=CCCCC=O
Glutardialdehyde Solution 25%, For Electron Microscopy, MilliporeSigma™
CAS: 111-30-8 Molecular Formula: C5H8O2 Molecular Weight (g/mol): 100.12 MDL Number: MFCD00007025 InChI Key: SXRSQZLOMIGNAQ-UHFFFAOYSA-N Synonym: glutaraldehyde,glutaral,glutaric dialdehyde,cidex,glutardialdehyde,1,5-pentanedial,sonacide,glutarol,aldesan,glutaric aldehyde PubChem CID: 3485 ChEBI: CHEBI:64276 IUPAC Name: pentanedial SMILES: O=CCCCC=O
StatLab Glutaraldehyde Buffer 2% pH 7.3
Glutaraldehyde Buffer 2% pH 7.3
Glutardialdehyde Solution 25%, For Electron Microscopy, acc. to P.J. Anderson, MilliporeSigma™
CAS: 111-30-8 Molecular Formula: C5H8O2 Molecular Weight (g/mol): 100.12 MDL Number: MFCD00007025 InChI Key: SXRSQZLOMIGNAQ-UHFFFAOYSA-N Synonym: glutaraldehyde,glutaral,glutaric dialdehyde,cidex,glutardialdehyde,1,5-pentanedial,sonacide,glutarol,aldesan,glutaric aldehyde PubChem CID: 3485 ChEBI: CHEBI:64276 IUPAC Name: pentanedial SMILES: O=CCCCC=O