missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ IPL-41 Insect Medium (1X)
GREENER_CHOICE

Catalog No. 11405081
Encompass
Change view
Click to view available options
Quantity:
1000 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11405081 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11405081 Supplier Gibco™ Supplier No. 11405081

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Requires

IPL-41 Insect Medium requires ambient CO2in order to maintain physiological pH.

A variety of options are available for a range of insect cell culture applications

Gibco IPL-41 Insect Medium was originally designed for the growth of Spodeptera frugiperda (Sf9) cells and the expression of recombinant Autographa Californica Nuclear Polyhedrosis Virus (rAcNPV) using the Baculovirus Expression Vector System (BEVS). This IPL-41 Insect Medium is modified as follows: With L-glutamine, Sodium bicarbonate. Without Tryptose phosphate broth and Yeastolate.The complete formulation is available. IPL-41 Insect Medium contains unique components required for insect cell culture, including malic acid, fumaric acid, and alpha-ketoglutaric acid. Gibco IPL-41 Insect Medium is commonly supplemented with 10% Fetal Bovine Serum (FBS) before use.

Cell Culture, High Five™ Cell Culture, Insect Cell Culture

Order Info

Shipping Condition: Room temperature

Specifications

Cell Line Sf21, Sf9
Cell Type Insect Cell
Form Liquid
Product Type Insect Cell Medium
With Additives Glutamine, Sodium Bicarbonate
Without Additives No Tryptose Phosphate Broth, No Yeastolate
Product Line Gibco
Quantity 1000 mL
Shipping Condition Room Temperature
Inorganic Salts Calcium
Content And Storage Storage conditions: 2-8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.