missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Glucose Solution

Catalog No. A2494001
Encompass
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2494001 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A2494001 Supplier Gibco™ Supplier No. A2494001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Gibco™ Glucose Solution contains D-glucose at a concentration of 200 g/L and is used as a serum-free medium supplement suitable for cell culture.

Gibco™ Glucose Solution contains D-glucose at a concentration of 200 g/L and is used as a serum-free medium supplement suitable for cell culture. Glucose Solution can be added to a variety of Gibco™ cell culture media including but not limited to Dulbecco’s Modified Eagle Medium (DMEM), RPMI 1640, DMEM/F-12, IMDM, and Minimum Essential Medium. Each lot of Glucose Solution is tested for sterility, osmolality, pH, and endotoxin level. We offer a variety of media to which Glucose Solution can be added. Find the right medium using the media selector tool.

Compliance

Glucose Solution is manufactured at a cGMP-compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

Specifications

Content And Storage Store in refrigerator (2–8°C) and protect from light.
Shelf life: 12 months from date of manufacture.
Description Glucose Solution, 50mL, Liquid, 12 Months Shelf Life, Used for Mammalian Cell Culture Applications, For Research Use Only
Classification Serum-free
Concentration 200 g/L
Form Liquid
Product Type Glucose
Sterility Sterile-filtered
For Use With (Application) Mammalian cell culture
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Product Line Gibco
Quantity 50 mL
Regulatory Status RUO
Show More Show Less
What is the glucose concentration in Glucose Solution (Cat. No. A249001)?

The glucose concentration of Glucose Solution (Cat. No. A2494001) is 200 grams of glucose per 1L of distilled water.

What solvent is used in the Glucose Solution (Cat. No. A2494001)?

The Glucose Solution (Cat. No. A2494001) solvent is distilled water.

For Research Use Only. Not for use in diagnostic procedures.