missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals G3335

CAS: 36099-95-3 | C16H19N3O5, C16H19N3O5 | 333.34 g/mol

Supplier:  Thermo Scientific Chemicals J64528MC

Quantity 100 mg

 Afficher plus de versions de ce produit

Numéro de catalogue. AAJ64528MC


Il en reste null

Description

Description

A cell-permeable dipeptide that acts as a selective and reversible peroxisome proliferator-activated receptor antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A cell-permeable dipeptide that acts as a selective and reversible peroxisome proliferator-activated receptor antagonist
Spécifications

Identifiants chimiques

36099-95-3
C16H19N3O5, C16H19N3O5
H-Trp-Glu-OH

Spécifications

100 mg
2°C to 8°C
36099-95-3
333.34g/mol
Light Sensitive
Solid
NC(CC1=CNC2=CC=CC=C12)C(=O)NC(CCC(O)=O)C(O)=O
333.34
White
A cell-permeable dipeptide that acts as a selective and reversible peroxisome proliferator-activated receptor antagonist
C16H19N3O5, C16H19N3O5
H-Trp-Glu-OH
Signal Transduction Reagent
PWIQCLSQVQBOQV-UHFFFAOYNA-N
2-[2-amino-3-(1H-indol-3-yl)propanamido]pentanedioic acid
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

Recommended Storage : Keep cold

FDS
Documents

Documents

Promotions

Promotions