missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formaldehyde Solution, 10% v/v, Reagents
Formaldehyde Solution, 10% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc CF1010004A
Spécifications
| Formaldehyde Solution, 10% v/v | |
| Colorless | |
| Poly Bottle | |
| 4 L | |
| Formalin | |
| Reagent |
| 7732-18-5,50-00-0 | |
| 100°C | |
| HCHO/H2O | |
| 1.01 | |
| 10% | |
| Liquid |