Learn More
Forchlorfenuron, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 456290050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spécifications
| Forchlorfenuron | |
| C12H10ClN3O | |
| Solubility in water: 0.039g/L. Other solubilities: soluble in methanol (119g/L), ethanol (149g/L),, acetone (127g/L);, slightly soluble in chloroform (2.7g/L) | |
| ClC1=CC(NC(=O)NC2=CC=CC=C2)=CC=N1 | |
| 247.68 | |
| CHEBI:81861 | |
| ≥97.5% (HPLC) |
| 68157-60-8 | |
| MFCD00059898 | |
| GPXLRLUVLMHHIK-UHFFFAOYSA-N | |
| 1-(2-chloropyridin-4-yl)-3-phenylurea | |
| 93379 | |
| 247.68 |
Identifiants chimiques
| 68157-60-8 | |
| 247.68 | |
| GPXLRLUVLMHHIK-UHFFFAOYSA-N | |
| CHEBI:81861 | |
| ClC1=CC(NC(=O)NC2=CC=CC=C2)=CC=N1 |
| C12H10ClN3O | |
| MFCD00059898 | |
| 93379 | |
| 1-(2-chloropyridin-4-yl)-3-phenylurea |
Sécurité et manipulation
GHS H Statement Suspected of causing cancer. Toxic to aquatic life with long lasting effects.
GHS P Statement Obtain special instructions before use. Use personal protective equipment as required. IF exposed or concerned: Get medical advice/attention. Avoid release to the environment.
Warning
RUO â Research Use Only