missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluoride Standard, 2ppm F- Premixed with Equal Volume of TISAB II, Certified, 2.00ppm ±0.05ppm, LabChem™
Supplier: LabChem LC146302
Specifications
| Fluoride Standard | |
| Poly Bottle | |
| 91.848,2.9,2.85,2.2,0.2 | |
| Liquid | |
| 2.00 ppm ±0.05 ppm | |
| MFCD00003524 | |
| Soluble in water | |
| [F-].[Na+] | |
| 41.99 | |
| 5235 | |
| 41.99 |
| 2ppm F- Premixed with Equal Volume of TISAB II | |
| 7681-49-4 , 7647-14-5 , 64-19-7 , 1310-73-2 , 125572-95-4 | |
| Passes Test | |
| FNa | |
| NaF | |
| Colorless | |
| PUZPDOWCWNUUKD-UHFFFAOYSA-M | |
| sodium fluoride | |
| 1 L | |
| CHEBI:28741 | |
| Certified |
Chemical Identifiers
| 7681-49-4 | |
| 41.99 | |
| PUZPDOWCWNUUKD-UHFFFAOYSA-M | |
| CHEBI:28741 | |
| [F-].[Na+] |
| FNa | |
| MFCD00003524 | |
| 5235 | |
| sodium fluoride |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature