missing translation for 'onlineSavingsMsg'
Learn More

Fluoride Standard, 2ppm F- Premixed with Equal Volume of TISAB II, Certified, 2.00ppm ±0.05ppm, LabChem™

Catalog No. LC146301
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC146301 500 mL Poly Bottle
LC146302 1 L Poly Bottle
LC146304 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC146301 Supplier LabChem Supplier No. LC146301

Chemical Identifiers

CAS 7681-49-4 , 7647-14-5 , 64-19-7 , 1310-73-2 , 125572-95-4
Molecular Formula FNa
Molecular Weight (g/mol) 41.99
MDL Number MFCD00003524
InChI Key PUZPDOWCWNUUKD-UHFFFAOYSA-M
PubChem CID 5235
ChEBI CHEBI:28741
IUPAC Name sodium fluoride
SMILES [F-].[Na+]

Specifications

Packaging Poly Bottle
CAS Max % 91.848, 2.9, 2.85, 2.2, 0.2
Identification Passes Test
Physical Form Liquid
Concentration 2.00 ppm ±0.05 ppm
Linear Formula NaF
Color Colorless
Solubility Soluble in water
Quantity 500 mL
Formula Weight 41.99
Grade Certified
Chemical Name or Material Fluoride Standard, 2ppm F- Premixed with Equal Volume of TISAB II
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature