missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluoride Standard, 1ppm F- Premixed with Equal Volume of TISAB II, Certified, 1.00ppm ±0.05ppm, LabChem™
$108.96 - $368.32
Chemical Identifiers
| CAS | 7681-49-4 |
|---|---|
| Molecular Formula | FNa |
| Molecular Weight (g/mol) | 41.99 |
| MDL Number | MFCD00003524 |
| InChI Key | PUZPDOWCWNUUKD-UHFFFAOYSA-M |
| PubChem CID | 5235 |
| ChEBI | CHEBI:28741 |
| IUPAC Name | sodium fluoride |
| SMILES | [F-].[Na+] |
Chemical Identifiers
| 7681-49-4 | |
| 41.99 | |
| PUZPDOWCWNUUKD-UHFFFAOYSA-M | |
| CHEBI:28741 | |
| [F-].[Na+] |
| FNa | |
| MFCD00003524 | |
| 5235 | |
| sodium fluoride |
Specifications
| Poly Bottle | |
| 91.849,2.9,2.85,2.2,0.2 | |
| Liquid | |
| 1.00 ppm ±0.05 ppm | |
| MFCD00003524 | |
| Soluble in water | |
| [F-].[Na+] | |
| 41.99 | |
| 5235 | |
| 41.99 | |
| Fluoride Standard, 1ppm F- Premixed with Equal Volume of TISAB II |
| 7681-49-4 , 7647-14-5 , 64-19-7 , 1310-73-2 , 125572-95-4 | |
| Passes Test | |
| FNa | |
| NaF | |
| Colorless | |
| PUZPDOWCWNUUKD-UHFFFAOYSA-M | |
| sodium fluoride | |
| 500 mL | |
| CHEBI:28741 | |
| Certified |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature