missing translation for 'onlineSavingsMsg'
Learn More

Fluorescamine

For fluorescent detection of amino acids, peptides and proteins | CAS: 38183-12-9 | C17H10O4 | 278.26 g/mol

Supplier:  Thermo Scientific Chemicals 043749MC

 Afficher plus de versions de ce produit

Numéro de catalogue. AA43749MC


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

For fluorescent detection of amino acids, peptides and proteins.Fluorescamine plays an important role as a reagent for the detection of amines, peptides and proteins. It is also used as a substrate for measuring protease activity. It is utilized in the detection and quantitation of residual aminopenicillins and sulfonamides in honey by HPLC. It finds application in the determination of lisinopril in human plasma and urine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Spécifications

Fluorescamine
155°C to 158°C
MFCD00005928
14,4158
Soluble in acetone,ethanol,chloroform,dimethylsulfoxide and acetonitrile. Slightly soluble in water.
O=C1OC2(OC=C(C2=O)C2=CC=CC=C2)C2=CC=CC=C12
278.26
278.27
38183-12-9
C17H10O4
100 mg
fluorescamine, fluram, spiro furan-2 3h ,1' 3'h-isobenzofuran-3,3'-dione, 4-phenyl, 4'-phenylspiro 2-benzofuran-3,2'-furan-1,3'-dione, 4-phenylspiro furan-2 3h ,1'-phthalan-3,3'-dione, 4-phenylspiro furan-2 3h-1'-3'-h-isobenzofuran-3,3'-dione, zfkjvjidpqddfy-uhfffaoysa, 4'-phenylspiro 2-benzofuran-3,2'-furan-1,3'-dion
ZFKJVJIDPQDDFY-UHFFFAOYNA-N
4'-phenylspiro[2-benzofuran-3,2'-furan]-1,3'-dione
37927
Powder

Identifiants chimiques

38183-12-9
278.26
ZFKJVJIDPQDDFY-UHFFFAOYNA-N
37927
C17H10O4
MFCD00005928
fluorescamine, fluram, spiro furan-2 3h ,1' 3'h-isobenzofuran-3,3'-dione, 4-phenyl, 4'-phenylspiro 2-benzofuran-3,2'-furan-1,3'-dione, 4-phenylspiro furan-2 3h ,1'-phthalan-3,3'-dione, 4-phenylspiro furan-2 3h-1'-3'-h-isobenzofuran-3,3'-dione, zfkjvjidpqddfy-uhfffaoysa, 4'-phenylspiro 2-benzofuran-3,2'-furan-1,3'-dion
O=C1OC2(OC=C(C2=O)C2=CC=CC=C2)C2=CC=CC=C12
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

missing translation for 'einecsNumber' : 253-814-5

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

FDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only