Learn More
Fisherbrand™ Pasteur Pipets
Being packaged in strong corrugated material provides maximum protection from external shock for Fisherbrand™ Pasteur Pipets. Each pack also contains a foam pad to protect the pipet tips.
 Afficher plus de versions de ce produit
Numéro de catalogue. 22-183-624
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
Being packaged in strong corrugated material provides maximum protection from external shock for Fisherbrand™ Pasteur Pipets. Each pack also contains a foam pad to protect the pipet tips.
  • Straight-cut tips and smooth, fire-polished tops to facilitate bulb attachment
  • Positive constriction approximately 1 in. from the top to prevent cotton plugs from slipping into pipet barrel
  • Easel configuration of each pack keeps pipets orderly and easily accessible
Spécifications
Spécifications
0.27 in.
Borosilicate Glass
4 packs of 250 in one box
Smooth fire polished
14.6 cm
7 mm
5.75 in.
Straight
2 mL
Pasteur Pipet
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS
Documents
Documents
Accessoires