Learn More
Epredia™ Fine Pore Cassettes
For very small biopsy tissue samples, cassettes have a front-edge lid hinge with attached lids that are available in 12 different color varieties
Supplier:  Epredia™ B851735TN
 Afficher plus de versions de ce produit
Numéro de catalogue. B851735TN
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
Printing
  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
  • Precision engineering for solid connection to microtomes
Archiving
  • Cassette surface texture optimizes scannability for archiving
Spécifications
Spécifications
Fine Pore
1000/Cs.
No
Cassette
Yes
Loose Packed
Tan
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS