missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ FGFR3 (G697C) (Human) Recombinant Protein

Numéro de catalogue. 89339383
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
89339383 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 89339383 Fournisseur Abnova™ Code fournisseur P6493
Il en reste null

For use with Functional Study.

Human FGFR3 (NP_000133.1, 436 a.a. - 806 a.a.) G697C mutant partial recombinant protein with GST-tag at N-terminal using baculovirus expression system.

Spécifications

For Use With (Application) Functional Study
Gene ID (Entrez) 2261
Name fibroblast growth factor receptor 3
Quality Control Testing The purity was assessed by SDS-PAGE/CBB staining.
Quantity 5 μg
Storage Requirements Stored at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias ACH, CD333, CEK2, HSFGFR3EX, JTK4