missing translation for 'onlineSavingsMsg'
Learn More

Ferric Ammonium Sulfate, For Chloride (Volhard's Indicator), Certified, 8%, LabChem™

$106.71 - $192.50

Identifiants chimiques

CAS 7783-83-7
Molecular Formula FeH28NO20S2
Molecular Weight (g/mol) 482.176
InChI Key LCPUDZUWZDSKMX-UHFFFAOYSA-K
PubChem CID 197097
IUPAC Name azanium;iron(3+);disulfate;dodecahydrate
SMILES [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
LC142701
Afficher les documents
LabChem
LC142701
500 mL Poly Bottle
chaque for $106.71
Il en reste null
 
LC142702
Afficher les documents
LabChem
LC142702
1 L Poly Bottle
chaque for $192.50
Il en reste null
 
Description

Description

Spécifications

Identifiants chimiques

7783-83-7
482.176
197097
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]
FeH28NO20S2
LCPUDZUWZDSKMX-UHFFFAOYSA-K
azanium;iron(3+);disulfate;dodecahydrate

Spécifications

7783-83-7,7732-18-5
FeH28NO20S2
Soluble in water
LCPUDZUWZDSKMX-UHFFFAOYSA-K
azanium;iron(3+);disulfate;dodecahydrate
197097
8%
Poly Bottle
Sulfur compounds; Gaseous Ammonia; Nitrogen oxides
500 mL
Ferric Ammonium Sulfate, For Chloride (Volhard's Indicator)
92,8
FeNH4S2O8·12H2O
Passes Test
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]
482.176
482.19
Certified
Passes Test
Brown/Yellow
Liquid
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

FDS
missing translation for 'documents'

missing translation for 'documents'