Learn More
Thermo Scientific™ RevertAid H Minus Reverse Transcriptase (200 U/μL)
Description
Thermo Scientific RevertAid H Minus Reverse Transcriptase is a recombinant M-MuLV RT. The enzyme possesses an RNA-dependent and DNA-dependent polymerase activity, but lacks RNase H activity due to a point mutation in the RNase H domain. RevertAid H Minus Reverse Transcriptase does not degrade RNA in RNA-DNA hybrids during synthesis of the first strand cDNA, and therefore high yields of full-length cDNA from long templates are obtained. RevertAid H Minus Reverse Transcriptase maintains activity over a wide temperature range (42–50°C).
Features of RevertAid H Minus Reverse Transcriptase include:
• High yields of full-length first strand cDNA up to 13 kb
• Optimum activity at 42–45°C
• Active up to 55°C
• Incorporates modified nucleotides (e.g., Cy3-, Cy5-, rhodamine-, aminoallyl-, and fluorescein-labeled nucleotides)
Applications
• First strand cDNA synthesis for RT-PCR and real-time RT-PCR
• Reverse transcription at elevated temperatures to reduce effects of secondary structure
• Synthesis of cDNA for cloning and expression
• Generation of labeled cDNA probes for microarrays
• DNA labeling
• Analysis of RNA by primer extension
Spécifications
Spécifications
| Concentration | 200 U/μL |
| Content And Storage | • RevertAid H Minus Reverse Transcriptase (200 U/μL) |
| Format | Stand-alone Enzyme |
| Reaction Speed | 60 min. |
| Technique | Reverse Transcription |
| Optimal Reaction Temperature | 42°C to 45°C |
| Quantity | 10,000 Units |
| Reverse Transcriptase | RevertAid H Minus |
| Ribonuclease H Activity | Reduced |
| Shipping Condition | Dry Ice |
| Afficher plus de résultats |
For Research Use Only. Not for use in diagnostic procedures.