missing translation for 'onlineSavingsMsg'
Learn More

Fehling's Solution 'B', Alkaline No. 2, Certified, LabChem™

Supplier:  LabChem LC142101

 Afficher plus de versions de ce produit

Numéro de catalogue. LC142101


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Spécifications

Spécifications

Fehling's Solution 'B', Alkaline No. 2
60.9,34.6,4.5
KNaC4H4O6·4H2O
Soluble in water
282.22
Poly Bottle
Sodium oxide; Carbon monoxide; Carbon dioxide; Thermal decomposition generates: Corrosive vapors
500 mL
Alkaline No. 2
6381-59-5,1310-73-2
UN1824
Passes Test
Certified
Passes Test
Colorless
Liquid
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions