missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl (Trimethylsilyl)acetate 93.0+%, TCI America™
Supplier: TCI America T158425G
Spécifications
| Ethyl (Trimethylsilyl)acetate | |
| Yellow | |
| C7H16O2Si | |
| 25 g | |
| ethyl trimethylsilyl acetate, ethyl trimethylsilylacetate, ethyl 2-trimethylsilyl acetate, etsa, trimethylsilyl acetic acid ethyl ester, acetic acid, trimethylsilyl-, ethyl ester, pubchem16058, acmc-1ahie | |
| CCOC(=O)C[Si](C)(C)C | |
| 160.288 | |
| 160.29 | |
| Liquid |
| 4071-88-9 | |
| 159°C | |
| MFCD00009172 | |
| 3272 | |
| QQFBQBDINHJDMN-UHFFFAOYSA-N | |
| ethyl 2-trimethylsilylacetate | |
| 77687 | |
| ≥93.0% (GC) |
Identifiants chimiques
| 4071-88-9 | |
| 160.288 | |
| QQFBQBDINHJDMN-UHFFFAOYSA-N | |
| 77687 | |
| CCOC(=O)C[Si](C)(C)C |
| C7H16O2Si | |
| MFCD00009172 | |
| ethyl trimethylsilyl acetate, ethyl trimethylsilylacetate, ethyl 2-trimethylsilyl acetate, etsa, trimethylsilyl acetic acid ethyl ester, acetic acid, trimethylsilyl-, ethyl ester, pubchem16058, acmc-1ahie | |
| ethyl 2-trimethylsilylacetate |
Sécurité et manipulation
missing translation for 'tsca' : No