missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ SNAP i.d. 2.0™ Protein Detection Systems for Western Blotting
Detection system with unique vacuum-driven technology and built-in flow distributor that actively drives reagents through membrane
Supplier: MilliporeSigma™ SNAP2MB1
Comprend: SNAP i.d. 2.0™ base, holding Frames, two antibody collection trays, vacuum tubing, rolling pad and blot roller, two wetting trays and quick start guide
Description
- System produces blots of high quality
- For use on immunoblots
- Detection Method: Chemiluminescence Chromogenic Fluorescence
- Mini and MultiBlot: 7.5 x 8.4cm and 4.5 x 8.4cm
- Midi and MultiBlot: 8.5 x 13.5cm and 4.5 x 8.4cm
Western Blotting
Identifiants chimiques
| Western Blot | |
| SNAP i.d. 2.0™ base, holding Frames, two antibody collection trays, vacuum tubing, rolling pad and blot roller, two wetting trays and quick start guide | |
| 1/Pk. |
Spécifications
| Western Blot | |
| SNAP i.d. 2.0™ base, holding Frames, two antibody collection trays, vacuum tubing, rolling pad and blot roller, two wetting trays and quick start guide | |
| 7.5 x 8.4 cm and 4.5 x 8.4 cm |
| Mini and MultiBlot | |
| 1/Pk. |