missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ SNAP i.d. 2.0™ Protein Detection Systems for Western Blotting

Detection system with unique vacuum-driven technology and built-in flow distributor that actively drives reagents through membrane

$2,840.00

Specifications

For Use With (Application) Western Blot
Includes SNAP i.d. 2.0™ base, holding Frames, two antibody collection trays, vacuum tubing, rolling pad and blot roller, two wetting trays and quick start guide
Quantity 1/Pk.
View More Specs

Includes: SNAP i.d. 2.0™ base, holding Frames, two antibody collection trays, vacuum tubing, rolling pad and blot roller, two wetting trays and quick start guide

Products 2
Catalog Number Mfr. No. Type Dimensions (L x W x H) Price Quantity & Availability  
Catalog Number Mfr. No. Type Dimensions (L x W x H) Price Quantity & Availability  
SNAP2MB2
View Documents Promotion Details
MilliporeSigma™
SNAP2MB2
Midi and MultiBlot 8.5 x 13.5 cm and 4.5 x 8.4 cm
Each for $2,840.00
Only null left
 
SNAP2MB1
View Documents Promotion Details
MilliporeSigma™
SNAP2MB1
Mini and MultiBlot 7.5 x 8.4 cm and 4.5 x 8.4 cm
Each for $2,840.00
Only null left
 
Description

Description

  • System produces blots of high quality
  • For use on immunoblots
  • Detection Method: Chemiluminescence Chromogenic Fluorescence
  • Mini and MultiBlot: 7.5 x 8.4cm and 4.5 x 8.4cm
  • Midi and MultiBlot: 8.5 x 13.5cm and 4.5 x 8.4cm

Western Blotting

Specifications

Specifications

Western Blot
SNAP i.d. 2.0™ base, holding Frames, two antibody collection trays, vacuum tubing, rolling pad and blot roller, two wetting trays and quick start guide
1/Pk.
Videos
SDS
Documents

Documents