missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Human ES/iPS Neural Induction Medium

Catalog No. SCM110
Change view
Click to view available options
Quantity:
125 mL/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCM110 125 mL/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCM110 Supplier MilliporeSigma™ Supplier No. SCM110

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

an serum free/chemically defined optimized media for the rapid production of expandable neural progenitor cells from pluripotent human ES and iPS cells.

Specifications

For Use With (Application) Cell Differentiation
Quantity 125 mL/Kit
Content And Storage -20°C