missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ EZ-Fit™ Filtration Unit
For bioburden testing of liquid samples like water, process samples, or final products
$512.00 - $616.55
Specifications
| Material (Membrane) | MCE |
|---|---|
| Pore Size | 0.45 μm |
| Sterility | Sterile |
| No. per Pack | 48 |
| Quantity | 48/Pk. |
| Catalog Number | Mfr. No. | Color | Volume (Metric) | Individually wrapped | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Color | Volume (Metric) | Individually wrapped | Price | Quantity & Availability | |||
|
EFHAB100B
|
MilliporeSigma™
EFHAB100B |
White with Black Grid | 100 mL | Bulk Packed |
Each for $512.00
|
|
|||
|
EFHAB100I
|
MilliporeSigma™
EFHAB100I |
White with Black Grid | 100 mL | Individually Wrapped |
Each for $539.00
|
|
|||
|
EFHAB250I
|
MilliporeSigma™
EFHAB250I |
White with Black Grid | 250 mL | Individually Wrapped |
Each for $602.00
|
|
|||
|
EFHAW100B
|
MilliporeSigma™
EFHAW100B |
Black with White Grid | 100 mL | Bulk Packed |
Each for $573.58
|
|
|||
|
EFHAW100I
|
MilliporeSigma™
EFHAW100I |
Black with White Grid | 100 mL | Individually Wrapped |
Pack of 48 Each for $576.62
|
|
|||
|
EFHAW250B
|
MilliporeSigma™
EFHAW250B |
Black with White Grid | 250 mL | Bulk Packed |
Each for $520.00
|
|
|||
|
EFHAW250I
|
MilliporeSigma™
EFHAW250I |
Black with White Grid | 250 mL | Individually Wrapped |
Each for $616.55
|
|
|||
Description
- Designed for optimizing and securing laboratory workflow to provide time-saving and reliable microbiological results
- Comprise of 100mL or 250mL funnels
- Black or white gridded membranes
- After filtration of sample, membrane can be transferred and cultured on agar plate
- Sterilized and disposable
- Units are either individually bagged or packaged in bulk
Conform to international standards (EP/USP) and comply with water testing regulations.
Specifications
| MCE | |
| Sterile | |
| 48/Pk. |
| 0.45 μm | |
| 48 | |
| Filtration Unit |