missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ EZ-Fit™ Filtration Unit
For bioburden testing of liquid samples like water, process samples, or final products
Supplier: MilliporeSigma™ EFHAB250I
Description
- Designed for optimizing and securing laboratory workflow to provide time-saving and reliable microbiological results
- Comprise of 100mL or 250mL funnels
- Black or white gridded membranes
- After filtration of sample, membrane can be transferred and cultured on agar plate
- Sterilized and disposable
Conform to international standards (EP/USP) and comply with water testing regulations.
Chemical Identifiers
| MCE | |
| Sterile | |
| 48/Pk. |
| 0.45 μm | |
| 48 | |
| Filtration Unit |
Specifications
| White with Black Grid | |
| 0.45 μm | |
| Individually Wrapped | |
| 48/Pk. | |
| 250 mL |
| MCE | |
| Sterile | |
| 48 | |
| Filtration Unit |