missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ EZ-Fit™ Filtration Unit

For bioburden testing of liquid samples like water, process samples, or final products

Supplier:  MilliporeSigma™ EFHAB100I

 View more versions of this product

Catalog No. EFHAB100I


Only null left

Description

Description

  • Designed for optimizing and securing laboratory workflow to provide time-saving and reliable microbiological results
  • Comprise of 100mL or 250mL funnels
  • Black or white gridded membranes
  • After filtration of sample, membrane can be transferred and cultured on agar plate
  • Sterilized and disposable

Conform to international standards (EP/USP) and comply with water testing regulations.

Specifications

Chemical Identifiers

MCE
Sterile
48/Pk.
0.45 μm
48
Filtration Unit

Specifications

White with Black Grid
0.45 μm
Individually Wrapped
48/Pk.
100 mL
MCE
Sterile
48
Filtration Unit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions