missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ ESGRO Complete™ PLUS Clonal Grade Medium

Catalog No. SF001100P
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SF001100P 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SF001100P Supplier MilliporeSigma™ Supplier No. SF001100P

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

a defined serum-free medium provided with a selective GSK3β inhibitor to enhance viability of mouse embryonic stem cells and increased maintenance of pluripotency in the absence of serum and feeder cells.

ESGRO Complete PLUS Clonal Grade Medium is a defined serum-free medium provided with a selective GSK3β inhibitor to enhance viability of mouse embryonic stem (ES) cells and increased maintenance of pluripotency in the absence of serum and feeder cells. This medium is intended for mouse ES cell expansion and growth at clonal density in serum-free conditions. The mouse ES cells should maintain germ line competency.

Specifications

For Use With (Application) Stem Cell Culture
Quantity 100 mL
Content And Storage Store in dark at -20° C