missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ ENStem-A™ Neural Expansion Medium

Catalog No. SCM004
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCM004 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCM004 Supplier MilliporeSigma™ Supplier No. SCM004

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

a defined serum-free formulation that has been optimized for the culture and expansion of ENStem-A Human Neural Progenitor Cells.

ENStem-A Neural Expansion Medium is a defined serum-free formulation that has been optimized for the culture and expansion of ENStem-A Human Neural Progenitor Cells. When used in conjunction with L-Glutamine (not provided) and FGF-2 (provided), the expansion medium will allow for the maintenance and proliferation of ENStem-ATM Human Neural Progenitor Cells.

Specifications

Classification Serum-free
For Use With (Application) Stem Cell Culture
Quantity 500 mL
Content And Storage −20°C