Learn More
National Optical™ Elementary Microscope with Tablet
Introducing the new line of digital compound microscopes with integrated 8 in. LCD tablet! The BTI1-205-LED features high speed, full-resolution imaging technology built into some of our most popular microscopes. This WiFi digital tablet transmits live images to iOS or Android devices. Tablet includes preloaded Motic apps and requires HDMI for live sharing.
Supplier: National Optical™ BTI1205LED
Description
Introducing the new line of digital compound microscopes with detachable LCD tablet! The BTI1-205-LED features high speed, full-resolution imaging technology built into some of our most popular microscopes.This WiFi digital tablet transmits live images to iOS or Android devices.Use it as a conventional microscope or share live images with colleagues using WiFi tablets, wireless laptops, and wireless devices. Tablet includes preloaded Motic apps. Connect, view, and share images easily and affordably with the BTI1-205-RLED.
OPTICAL SYSTEM
- Widefield 10x /18 mm eyepiece
- 30° inclined forward-facing monocular head
- 4X, 10X and 40X Achromatic objectives
ILLUMINATION
- 70 mW/3.5 V rechargeable LED illumination with intensity control
FOCUSING
- Separate coarse and fine focus
STAGE
- Floating and rotating stage with unique one-touch stage clips
FRAME
- Rugged cast aluminum, gray enamel finish
TABLET
- 8 in. LCD Android tablet with 1280 x 800 screen resolution
- Connection: WiFi; micro SD card
- WiFi Resolution 1920 x 1080 / 1280 x 720
- 4.0 MP capture resolution
- WiFi (2.4 or 5GHz) and Ethernet (RJ45)
- MotiConnect App
- Connect 5 WiFi devices at one time
- Connect office/clinic/business network through RJ45 port connection
PACKING
- Molded styrofoam container
- Tablet shipped in same box with microscope
- Individual: Ship weight 8.2 kg (18 lb.)
- Dimensions: 28 x 23 x 64 cm (11 x 9 x 25 in.)
- Master carton pack 1 each: Ship weight: 8.2 kg (18 lb.), dimensions 28 x 23 x 64 cm (11 x 9 x 25 in.)
Spécifications
| WF 10X/18 mm | |
| LED | |
| DIN Achromatic | |
| 110/220 V |
| Monocular | |
| 4X, 10X, 40X | |
| Beginner Microscope |