missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDTA disodium salt solution, Volumetric, 0.05Â M EDTA-Na2, Honeywell Fluka™
Volumetric, 0.05Â M EDTA-Na2
$89.18 - $89.18
Identifiants chimiques
| CAS | 139-33-3 |
|---|---|
| Molecular Formula | C10H14N2Na2O8 |
| Molecular Weight (g/mol) | 336.21 |
| MDL Number | MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037 |
| InChI Key | ZGTMUACCHSMWAC-UHFFFAOYSA-L |
| Synonym | ethylenediaminetetraacetic acid disodium salt, edta na2, disodium 2-2-bis carboxymethyl amino ethyl-carboxymethyl amino acetic acid |
| PubChem CID | 57339238 |
| ChEBI | CHEBI:64734 |
| SMILES | [Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|
6002395
|
Honeywell Chemicals
318892500ML |
500 mL |
chaque for $89.18
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Identifiants chimiques
| 139-33-3 | |
| 336.21 | |
| ZGTMUACCHSMWAC-UHFFFAOYSA-L | |
| 57339238 | |
| [Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O |
| C10H14N2Na2O8 | |
| MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037 | |
| ethylenediaminetetraacetic acid disodium salt, edta na2, disodium 2-2-bis carboxymethyl amino ethyl-carboxymethyl amino acetic acid | |
| CHEBI:64734 |
Spécifications
| 139-33-3 | |
| C10H14N2Na2O8 | |
| MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037 | |
| 3822669 | |
| ZGTMUACCHSMWAC-UHFFFAOYSA-L | |
| disodium;2-[2-[bis(carboxylatomethyl)azaniumyl]ethyl-(carboxylatomethyl)azaniumyl]acetate | |
| 57339238 | |
| 336.21g/mol | |
| Ethylenediaminetetraacetic Acid Disodium Salt Solution |
| 500 mL | |
| [-CH2N(CH2CO2Na)CH2CO2H]2 | |
| NONH for all modes of transport | |
| ethylenediaminetetraacetic acid disodium salt, edta na2, disodium 2-2-bis carboxymethyl amino ethyl-carboxymethyl amino acetic acid | |
| [Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O | |
| 336.21 | |
| CHEBI:64734 | |
| 1.01g/mL at 25°C |