missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ IsoClean™ Coverall (all)
Tyvek coveralls provide an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash. Bound seams reduce the potential for particle penetration.
Supplier: DuPont™ IC253BWHXL00250B
Avertissement:
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Description
- For use in controlled environments with high standards for particle and microbiological contamination control:
- Pharmaceutical
- Biotech
- Medical Device
- Electronics
- Light and durable
- Spun polyethylene creates barrier to particles, micro-organisms, and non-hazardous liquids
- Lowest level particle shedding
- Bound seams and neck
- Covered elastic wrists and ankles with dolman sleeves
- Zipper closure
- Should not be worn around open flame, heat source, sparks or ignition sources
Spécifications
| Cleanroom Coveralls | |
| Unisex | |
| White | |
| Covered Elastic | |
| Collared | |
| Bound | |
| Bound | |
| 33.5 in. | |
| 85 cm | |
| 5.4 mil | |
| 38.25 in. | |
| 5.11 to 6.3 ft. | |
| Bulk |
| X-Large | |
| Tyvek | |
| Single-use | |
| Covered Elastic | |
| Zipper | |
| Non-sterile | |
| 40 to 43 in. | |
| 101.6 to 109.22 cm | |
| Dolman | |
| Barrier to Dry Particles, Microorganisms, and Non-Hazardous Liquids in Controlled Environments | |
| 97.15 cm | |
| 155.75 to 192.04 cm | |
| Class 1000 (ISO 6) |