missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ ProShield™ 10 Lab Coat
SMS disposable lab coats are designed for a range of industries for nonhazardous dry particle and light liquid splash applications. Available in blue or white in a variety of sizes.
Supplier: DuPont™ PB219SWHMD003000
Description
- Knit collar
- Set sleeve
- Knit cuff
- Manufactured under specifications that do not contain natural rubber latex
Spécifications
| General Purpose Lab Coat | |
| Unisex | |
| White | |
| Knit | |
| Non-sterile | |
| Knit | |
| PB219 | |
| ASTM D1777 |
| Medium | |
| ProShield | |
| Snaps | |
| Single-use | |
| 3 | |
| Yes | |
| Serged |