missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ ProShield™ 10 Hooded Coverall
Spunbond meltblown spunbond (SMS) coveralls protect against non-hazardous particles and light liquid splashes in a variety of industries.
Supplier: DuPont™ PB122SWH3X002500
Description
Spunbond meltblown spunbond (SMS) coveralls protect against non-hazardous particles and light liquid splashes in a variety of industries.
- Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
- Attached hood with elastic around face opening.
- Elastic opening for tighter fit at wrist
- Attached skid-resistant boots
- Manufactured under specifications that do not contain natural rubber latex
Spécifications
| General Purpose Coveralls | |
| Unisex | |
| White | |
| Elastic | |
| Elastic | |
| Attached | |
| Serged | |
| ASTM D1117 | |
| General Maintenance, Janitorial, Cleaning |
| 3X-Large | |
| ProShield | |
| Single-use | |
| Attached Boots | |
| Hooded | |
| Front Zipper | |
| Non-sterile | |
| PB122S | |
| 25/Cs. |