missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl Methylmalonate 98.0+%, TCI America™
Supplier: TCI America M0366100ML
Spécifications
| Diethyl Methylmalonate | |
| Colorless | |
| C8H14O4 | |
| 100 mL | |
| UPQZOUHVTJNGFK-UHFFFAOYSA-N | |
| diethyl 2-methylpropanedioate | |
| 11857 | |
| ≥98.0% (GC) |
| 609-08-5 | |
| 199°C | |
| MFCD00009162 | |
| diethyl methylmalonate, methylmalonic acid diethyl ester, diethyl 2-methylmalonate, propanedioic acid, methyl-, diethyl ester, 1,3-diethyl 2-methylpropanedioate, diethylmethylmalonate, ethyl methylmalonate, diethyl methylpropanedioate, malonic acid, methyl-, diethyl ester, 2-methylmalonic acid diethyl | |
| CCOC(=O)C(C)C(=O)OCC | |
| 174.196 | |
| 174.20 | |
| Liquid |
Identifiants chimiques
| 609-08-5 | |
| 174.196 | |
| UPQZOUHVTJNGFK-UHFFFAOYSA-N | |
| 11857 | |
| CCOC(=O)C(C)C(=O)OCC |
| C8H14O4 | |
| MFCD00009162 | |
| diethyl methylmalonate, methylmalonic acid diethyl ester, diethyl 2-methylmalonate, propanedioic acid, methyl-, diethyl ester, 1,3-diethyl 2-methylpropanedioate, diethylmethylmalonate, ethyl methylmalonate, diethyl methylpropanedioate, malonic acid, methyl-, diethyl ester, 2-methylmalonic acid diethyl | |
| diethyl 2-methylpropanedioate |
Sécurité et manipulation
TSCA : Yes