Learn More
Dichlorobis(di-tert-butylphenylphosphine)palladium(II), Pd 16.1%
CAS: 34409-44-4 | C28H46Cl2P2Pd | 621.94 g/mol
$102.22 - $310.70
Identifiants chimiques
| CAS | 34409-44-4 |
|---|---|
| Molecular Formula | C28H46Cl2P2Pd |
| Molecular Weight (g/mol) | 621.94 |
| MDL Number | MFCD09953450 |
| InChI Key | DFKFLLBQLYGVIH-UHFFFAOYSA-L |
| Synonym | dichlorobis di-tert-butylphenylphosphine palladium ii, bis di-t-butyl phenyl phosphine palladium dichloride, bis di-tert-butyl phenyl phosphane ; palladium chloride |
| PubChem CID | 56649816 |
| IUPAC Name | ditert-butyl(phenyl)phosphane;dichloropalladium |
| SMILES | Cl[Pd++]Cl.CC(C)(C)P(C1=CC=CC=C1)C(C)(C)C.CC(C)(C)P(C1=CC=CC=C1)C(C)(C)C |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|
AA45453MD
|
Thermo Scientific Chemicals
045453MD |
250 mg |
chaque for $102.22
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
AA4545303
|
Thermo Scientific Chemicals
04545303 |
1 g |
chaque for $310.70
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
Dichlorobis(di-tert-butylphenylphosphine)palladium(II) is used as an effective catalyst for cross-coupling reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
| 34409-44-4 | |
| 621.94 | |
| DFKFLLBQLYGVIH-UHFFFAOYSA-L | |
| 56649816 | |
| Cl[Pd++]Cl.CC(C)(C)P(C1=CC=CC=C1)C(C)(C)C.CC(C)(C)P(C1=CC=CC=C1)C(C)(C)C |
| C28H46Cl2P2Pd | |
| MFCD09953450 | |
| dichlorobis di-tert-butylphenylphosphine palladium ii, bis di-t-butyl phenyl phosphine palladium dichloride, bis di-tert-butyl phenyl phosphane ; palladium chloride | |
| ditert-butyl(phenyl)phosphane;dichloropalladium |
Spécifications
| 34409-44-4 | |
| C28H46Cl2P2Pd | |
| 250 mg | |
| dichlorobis di-tert-butylphenylphosphine palladium ii, bis di-t-butyl phenyl phosphine palladium dichloride, bis di-tert-butyl phenyl phosphane ; palladium chloride | |
| DFKFLLBQLYGVIH-UHFFFAOYSA-L | |
| ditert-butyl(phenyl)phosphane;dichloropalladium | |
| 56649816 | |
| Solid |
| (decomposition) | |
| MFCD09953450 | |
| Air and moisture sensitive | |
| Soluble in water and polar organic solvents. | |
| Cl[Pd++]Cl.CC(C)(C)P(C1=CC=CC=C1)C(C)(C)C.CC(C)(C)P(C1=CC=CC=C1)C(C)(C)C | |
| 621.94 | |
| 621.95 | |
| Dichlorobis(di-tert-butylphenylphosphine)palladium(II) |
Sécurité et manipulation
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only