Learn More
Thermo Scientific™ Deionizer Cartridges
Choose from an array of Thermo Scientific™ Deionizer Cartridges for select Thermo Scientific™ Barnstead™ water systems, based on your application.
$570.56 - $948.91
| Catalog Number | Mfr. No. | For Use With (Equipment) | Resin Type | Capacity | For Use With (Application) | Price | Quantity & Availability | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | For Use With (Equipment) | Resin Type | Capacity | For Use With (Application) | Price | Quantity & Availability | ||
|
090343
|
Thermo Scientific™
D 0809 |
Bantam™ Deionizer, B-Pure™ | Mixed Bed | 875 grains |
Each for $732.29
|
|
|||
|
090348
|
Thermo Scientific™
D 0811 |
B-Pure™ | Porous Strong Ion Exchange | 30 g | Oxygen Removal |
Each for $756.68
|
|
||
|
090345
|
Thermo Scientific™
D 0813 |
Bantam™ Deionizer, B-Pure™ | Activated Carbon | 5000 gal. | Organic Removal |
Each for $585.82
|
|
||
|
0903410
|
Thermo Scientific™
D 0815 |
Bantam™ Deionizer, B-Pure™ | Strong Cation Exchange | 3170 grains | Cation Removal |
Each for $570.56
|
|
||
|
0902833
|
Thermo Scientific™
D0832 |
Bantam™ Deionizer, B-Pure™ | Mixed Bed, Activated Carbon | 785 grains/1000 gal. | Ultrapure and Organic Removal |
Each for $646.85
|
|
||
|
09034104
|
Thermo Scientific™
D 0835 |
B-Pure™ | Macroreticular, Activated Carbon | 5000 gal. | Pretreatment |
Each for $692.62
|
|
||
|
090345C
|
Thermo Scientific™
D 0836 |
B-Pure™ | Macroreticular, Activated Carbon | 2000 gal. | Pretreatment Macropure |
Each for $948.91
|
|
||
|
0905047
|
Thermo Scientific™
D63112 |
B-Pure™ | Extruded Carbon | 5000 gal. | Organic Removal |
Each for $749.18
|
|
||
|
09050263
|
Thermo Scientific™
D 8809 |
B-Pure™ | Ultrapure, Oxygen Removal | 455 grains/30 g | Ultrapure and Organic Removal |
Each for $765.71
|
|
||
|
0903490
|
Thermo Scientific™
D8811 |
B-Pure™ | Ultrapure, Oxygen and Organic Removal | 365 grains/2000 gal./12 g | Ultrapure, Oxygen and Organic Removal |
Each for $649.00
|
|
||
|
09034
|
Thermo Scientific™
D 0803 |
Thermo Scientific™ Barnstead™ Bantam™ B-Pure™ or Nanopure™ Water Purification Systems as Indicated | Two Bed Ion Exchange | 1760 grains | High capacity, two-bed ion exchange |
Each for $698.71
|
|
||
Description
Choose from an array of Thermo Scientific™ Deionizer Cartridges based on your application needs. These cartridges are for use with the Thermo Scientific™ Barnstead™ Bantam™, B-Pure™ or Nanopure™ water purification systems as indicated.
- Choice of full or half cartridges
Specifications
| Deionizer Cartridge |