Learn More
Thermo Scientific™ Deionizer Cartridges
Choose from an array of Thermo Scientific™ Deionizer Cartridges for select Thermo Scientific™ Barnstead™ water systems, based on your application.
Supplier:  Thermo Scientific™ D0832
 Afficher plus de versions de ce produit
Numéro de catalogue. 0902833
Il en reste null
Description
Description
Choose from an array of Thermo Scientific™ Deionizer Cartridges based on your application needs. These cartridges are for use with the Thermo Scientific™ Barnstead™ Bantam™, B-Pure™ or Nanopure™ water purification systems as indicated.
  • Choice of full or half cartridges
Spécifications
Identifiants chimiques
Deionizer Cartridge
Spécifications
Cartridge for ultrapure and organic removal; mixed-bed, activated carbon; full size
Mixed Bed, Activated Carbon
Bantam™ Deionizer, B-Pure™
785 grains/1000 gal.
Ultrapure and Organic Removal
Deionizer Cartridge
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS