Learn More
Thermo Scientific™ Deionizer Cartridges
Catalog No. 090343
Change view
Click to view available options
For Use With (Equipment):
B-Pure™
Bantam™ Deionizer, B-Pure™
Thermo Scientific™ Barnstead™ Bantam™ B-Pure™ or Nanopure™ Water Purification Systems as Indicated
Resin Type:
Activated Carbon
Extruded Carbon
Macroreticular, Activated Carbon
Mixed Bed
Mixed Bed, Activated Carbon
Porous Strong Ion Exchange
Strong Cation Exchange
Two Bed Ion Exchange
Ultrapure, Oxygen and Organic Removal
Ultrapure, Oxygen Removal
Capacity:
1760 grains
3170 grains
365 grains/2000 gal./12 g
455 grains/30 g
785 grains/1000 gal.
875 grains
30 g
2000 gal.
5000 gal.
For Use With (Application):
Cation Removal
High capacity, two-bed ion exchange
Organic Removal
Oxygen Removal
Pretreatment
Pretreatment Macropure
Ultrapure
Ultrapure and Organic Removal
Ultrapure, Oxygen and Organic Removal
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Resin Type Capacity For Use With (Application)
090343 Bantam™ Deionizer, B-Pure™ Mixed Bed 875 grains -
1116457 B-Pure™ Mixed Bed - Ultrapure
090348 B-Pure™ Porous Strong Ion Exchange 30 g Oxygen Removal
090345 Bantam™ Deionizer, B-Pure™ Activated Carbon 5000 gal. Organic Removal
0903410 Bantam™ Deionizer, B-Pure™ Strong Cation Exchange 3170 grains Cation Removal
0902833 Bantam™ Deionizer, B-Pure™ Mixed Bed, Activated Carbon 785 grains/1000 gal. Ultrapure and Organic Removal
09034104 B-Pure™ Macroreticular, Activated Carbon 5000 gal. Pretreatment
090345C B-Pure™ Macroreticular, Activated Carbon 2000 gal. Pretreatment Macropure
0905047 B-Pure™ Extruded Carbon 5000 gal. Organic Removal
09050263 B-Pure™ Ultrapure, Oxygen Removal 455 grains/30 g Ultrapure and Organic Removal
0903490 B-Pure™ Ultrapure, Oxygen and Organic Removal 365 grains/2000 gal./12 g Ultrapure, Oxygen and Organic Removal
09034 Thermo Scientific™ Barnstead™ Bantam™ B-Pure™ or Nanopure™ Water Purification Systems as Indicated Two Bed Ion Exchange 1760 grains High capacity, two-bed ion exchange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 090343 Supplier Thermo Scientific™ Supplier No. D 0809
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Choose from an array of Thermo Scientific™ Deionizer Cartridges for select Thermo Scientific™ Barnstead™ water systems, based on your application.
Choose from an array of Thermo Scientific™ Deionizer Cartridges based on your application needs. These cartridges are for use with the Thermo Scientific™ Barnstead™ Bantam™, B-Pure™ or Nanopure™ water purification systems as indicated.
  • Choice of full or half cartridges
Specifications
Capacity 875 grains
Resin Type Mixed Bed
For Use With (Equipment) Bantam™ Deionizer, B-Pure™
Type Deionizer Cartridge