Learn More
- Home
- Products
- Water Purification
- Water Purification Filters and Cartridges
- Thermo Scientific™ Deionizer Cartridges
Thermo Scientific™ Deionizer Cartridges
Catalog No. 0905047
Change view
Click to view available options
For Use With (Equipment):
B-Pure™
Bantam™ Deionizer, B-Pure™
Thermo Scientific™ Barnstead™ Bantam™ B-Pure™ or Nanopure™ Water Purification Systems as Indicated
Resin Type:
Activated Carbon
Extruded Carbon
Macroreticular, Activated Carbon
Mixed Bed
Mixed Bed, Activated Carbon
Porous Strong Ion Exchange
Strong Cation Exchange
Two Bed Ion Exchange
Ultrapure, Oxygen and Organic Removal
Ultrapure, Oxygen Removal
Capacity:
1760 grains
3170 grains
365 grains/2000 gal./12 g
455 grains/30 g
785 grains/1000 gal.
875 grains
30 g
2000 gal.
5000 gal.
For Use With (Application):
Cation Removal
High capacity, two-bed ion exchange
Organic Removal
Oxygen Removal
Pretreatment
Pretreatment Macropure
Ultrapure and Organic Removal
Ultrapure, Oxygen and Organic Removal
11 product options available for selection
| Catalog No. | For Use With (Equipment) | Resin Type | Capacity | For Use With (Application) |
|---|---|---|---|---|
| 0905047 | B-Pure™ | Extruded Carbon | 5000 gal. | Organic Removal |
| 090343 | Bantam™ Deionizer, B-Pure™ | Mixed Bed | 875 grains | - |
| 090348 | B-Pure™ | Porous Strong Ion Exchange | 30 g | Oxygen Removal |
| 090345 | Bantam™ Deionizer, B-Pure™ | Activated Carbon | 5000 gal. | Organic Removal |
| 0903410 | Bantam™ Deionizer, B-Pure™ | Strong Cation Exchange | 3170 grains | Cation Removal |
| 0902833 | Bantam™ Deionizer, B-Pure™ | Mixed Bed, Activated Carbon | 785 grains/1000 gal. | Ultrapure and Organic Removal |
| 09034104 | B-Pure™ | Macroreticular, Activated Carbon | 5000 gal. | Pretreatment |
| 090345C | B-Pure™ | Macroreticular, Activated Carbon | 2000 gal. | Pretreatment Macropure |
| 09050263 | B-Pure™ | Ultrapure, Oxygen Removal | 455 grains/30 g | Ultrapure and Organic Removal |
| 0903490 | B-Pure™ | Ultrapure, Oxygen and Organic Removal | 365 grains/2000 gal./12 g | Ultrapure, Oxygen and Organic Removal |
| 09034 | Thermo Scientific™ Barnstead™ Bantam™ B-Pure™ or Nanopure™ Water Purification Systems as Indicated | Two Bed Ion Exchange | 1760 grains | High capacity, two-bed ion exchange |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
Description
Choose from an array of Thermo Scientific™ Deionizer Cartridges based on your application needs. These cartridges are for use with the Thermo Scientific™ Barnstead™ Bantam™, B-Pure™ or Nanopure™ water purification systems as indicated.
- Choice of full or half cartridges
Specifications
Specifications
| Capacity | 5000 gal. |
| Resin Type | Extruded Carbon |
| For Use With (Application) | Organic Removal |
| For Use With (Equipment) | B-Pure™ |
| Type | Deionizer Cartridge |