Learn More
Thermo Scientific™ Deionizer Cartridges
Choose from an array of Thermo Scientific™ Deionizer Cartridges for select Thermo Scientific™ Barnstead™ water systems, based on your application.
Supplier: Thermo Scientific™ D 0815
Description
Choose from an array of Thermo Scientific™ Deionizer Cartridges based on your application needs. These cartridges are for use with the Thermo Scientific™ Barnstead™ Bantam™, B-Pure™ or Nanopure™ water purification systems as indicated.
- Choice of full or half cartridges
Identifiants chimiques
| Deionizer Cartridge |
Spécifications
| Cartridge for cation removal; strong cation exchange; full | |
| Strong Cation Exchange | |
| Bantam™ Deionizer, B-Pure™ |
| 3170 grains | |
| Cation Removal | |
| Deionizer Cartridge |