Learn More
Decyl aldehyde, 95%
CAS: 112-31-2 | C10H20O | 156.27 g/mol
$54.08 - $54.08
Identifiants chimiques
| CAS | 112-31-2 |
|---|---|
| Molecular Formula | C10H20O |
| Molecular Weight (g/mol) | 156.27 |
| MDL Number | MFCD00007031 |
| InChI Key | KSMVZQYAVGTKIV-UHFFFAOYSA-N |
| Synonym | decyl aldehyde, capraldehyde, caprinaldehyde, decanaldehyde, n-decyl aldehyde, capric aldehyde, 1-decanal, n-decanal, n-decaldehyde, 1-decyl aldehyde |
| PubChem CID | 8175 |
| ChEBI | CHEBI:31457 |
| IUPAC Name | decanal |
| SMILES | CCCCCCCCCC=O |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|
AC154971000
|
Thermo Scientific Chemicals
154971000 |
100 mL | Glass bottle |
chaque for $54.08
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
| 112-31-2 | |
| 156.27 | |
| KSMVZQYAVGTKIV-UHFFFAOYSA-N | |
| 8175 | |
| decanal |
| C10H20O | |
| MFCD00007031 | |
| decyl aldehyde, capraldehyde, caprinaldehyde, decanaldehyde, n-decyl aldehyde, capric aldehyde, 1-decanal, n-decanal, n-decaldehyde, 1-decyl aldehyde | |
| CHEBI:31457 | |
| CCCCCCCCCC=O |
Spécifications
| 112-31-2 | |
| Colorless to Yellow | |
| 207.0°C to 209.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4270 to 1.4300 | |
| MFCD00007031 | |
| 01,711 | |
| decyl aldehyde, capraldehyde, caprinaldehyde, decanaldehyde, n-decyl aldehyde, capric aldehyde, 1-decanal, n-decanal, n-decaldehyde, 1-decyl aldehyde | |
| KSMVZQYAVGTKIV-UHFFFAOYSA-N | |
| decanal | |
| 8175 | |
| 156.27 | |
| Liquid |
| 7.0°C | |
| 0.8300g/mL | |
| 85°C | |
| 94% min. (GC) | |
| C10H20O | |
| CH3(CH2)8CHO | |
| 100 mL | |
| 0.83 | |
| Solubility in water: insoluble. Other solubilities: soluble in 80% alcohol,insoluble in glycerol | |
| CCCCCCCCCC=O | |
| 156.27 | |
| CHEBI:31457 | |
| 95% | |
| Decyl aldehyde, 95% |
Sécurité et manipulation
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 203-957-4
missing translation for 'rtecsNumber' : HD6000000
missing translation for 'tsca' : TSCA
RUO – Research Use Only