missing translation for 'onlineSavingsMsg'
Learn More

Decon™ Contrex™ AL Alkaline Liquid Detergent
GREENER_CHOICE

For use in manual, soak, or ultrasonic cleaning

$75.20 - $237.43

Spécifications

Product Type Alkaline Detergent
Color-coded No
Form Concentrated Liquid
pH 7.5 to 9.5
Scent Regular Scent
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Volume (English) Prix Quantity & Availability  
Numéro de catalogue Numéro du manufacturier. Volume (English) Prix Quantity & Availability  
0435810
GREENER_CHOICE
Afficher les documents Détails de la promotion
Decon™
5301
1 gal.
chaque for $237.43
caisse de 4 chaque for $949.72
Il en reste null
 
0435811
GREENER_CHOICE
Afficher les documents Détails de la promotion
Decon™
DC5332
1 qt.
chaque for $75.20
caisse de 12 chaque for $902.40
Il en reste null
 
Description

Description

For use in manual, soak, or ultrasonic cleaning

Ideal for cleaning labware, scientific apparatus, and process equipment.

  • Concentrated liquid
  • Free-rinsing, phosphate-free, and disposable straight to drain
  • Environmentally Friendly: Item contains environmental attributes or features (e.g. water conserving, lead-free, etc.) as described in the item detail
  • Recycled: Item contains recycled content as described in the item detail
  • Non-toxic: Item is non-toxic
Spécifications

Spécifications

Alkaline Detergent
Concentrated Liquid
Regular Scent
29 CFR1910.1200
Liquid
No
7.5 to 9.5
Non-sterile
Translucent Yellow
For use in manual, soak or ultrasonic cleaning of labware, scientific apparatus and process equipment
Videos
FDS
Documents

Documents