missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ DBI-Sala™ ShockWave 2™ Shock-Absorbing Lanyard
Unique stretchable design offers complete freedom of movement while reducing trip-fall hazards and dragging or snagging by expanding to 6 ft. (1.8 m) when taut and automatically contracting to 4-1/2 ft. (1.4 m) in reaction to the workerâs movements.
$184.10 - $184.10
Spécifications
| Arresting Force (English) | Energy Absorber Class: 6 ft (1.8 m) Free Fall/MAAF 900 lb. (4 kN) |
|---|---|
| Color | Yellow |
| Length (English) | 6 ft. |
| Length (Metric) | 1.8 m |
| Material | Zinc-plated Steel |
| Numéro de catalogue | Numéro du manufacturier. | Connector Types | Product Type | Prix | Quantité | ||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Connector Types | Product Type | Prix | Quantité | ||||
|
19-809-959
|
3M™
7100157035 |
Snap Hook | 6 ft. (1.8 m) single-leg with tubular web and snap hooks at each end |
chaque for $184.10
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
Description
Unique stretchable design allows complete freedom of movement, practically eliminating snagging and dragging.
- Stretches from to 4 1/2 to 6 ft. (1.3 to 1.8m) to reduce trip and fall hazards
- In the event of a fall, the inner core limits the fall arrest force to 900 lb. (408kg) or less, stopping the fall in a controlled, smooth manner
- After a fall, a stitch rips to reveal a hidden warning label that indicates the lanyard has had a load placed on it and must be replaced
- Durable, abrasion-resistant nylon outer webbing is rated at a minimum 6000 lb. (2722kg) of tensile strength
- Maximum capacity is 310 lb. (140kg)
Order Info
Available in a variety of styles.
Compliance
Meets ANSI™ Z359.7-2011 requirements.
Spécifications
| Energy Absorber Class: 6 ft (1.8 m) Free Fall/MAAF 900 lb. (4 kN) | |
| 6 ft. | |
| Zinc-plated Steel | |
| 130 to 310 lb. |
| Yellow | |
| 1.8 m | |
| Single-Leg | |
| Polyester Tubular Web |