missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Sorbitol, 97%

CAS: 50-70-4 | C6H14O6 | 182.17 g/mol

$51.77 - $530.53

Chemical Identifiers

CAS 50-70-4
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.17
MDL Number MFCD00004708
InChI Key FBPFZTCFMRRESA-JGWLITMVSA-N
Synonym d-sorbitol, sorbitol, d-glucitol, glucitol, l-gulitol, --sorbitol, diakarmon, glucarine, sorbol, d---sorbitol
PubChem CID 5780
ChEBI CHEBI:17924
IUPAC Name (2R,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol
SMILES C(C(C(C(C(CO)O)O)O)O)O
View More Specs

Products 6
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC132730050
View Documents Promotion Details
Thermo Scientific Chemicals
132730050
5 g Glass bottle
Each for $51.77
Only null left
 
AC132731000
View Documents Promotion Details
Thermo Scientific Chemicals
132731000
100 g Plastic bottle
Each for $53.61
Only null left
 
AC132735000
View Documents Promotion Details
Thermo Scientific Chemicals
132735000
500 g Plastic bottle
Each for $63.80
Only null left
 
AC132730010
View Documents Promotion Details
Thermo Scientific Chemicals
132730010
1 kg Plastic bottle
Each for $101.63
Only null left
 
AC132730025
View Documents Promotion Details
Thermo Scientific Chemicals
132730025
2.5 kg Plastic bottle
Each for $214.15
Only null left
 
AC132730100
View Documents Promotion Details
Thermo Scientific Chemicals
132730100
10 kg Plastic drum
Each for $530.53
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

50-70-4
182.17
FBPFZTCFMRRESA-JGWLITMVSA-N
5780
(2R,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol
C6H14O6
MFCD00004708
d-sorbitol, sorbitol, d-glucitol, glucitol, l-gulitol, --sorbitol, diakarmon, glucarine, sorbol, d---sorbitol
CHEBI:17924
C(C(C(C(C(CO)O)O)O)O)O

Specifications

50-70-4
White
96% min. (on anhydrous substance) (HPLC)
MFCD00004708
158851
+ 4.00
Solubility in water: soluble. Other solubilities: slightly soluble in acetic acid, soluble in glycerol and propylene glycol, slightly soluble in methanol, ethanol, phenol,, acetamide, acetone, dmf and pyridine, practically insoluble in most other org. sol
C(C(C(C(C(CO)O)O)O)O)O
182.17
CHEBI:17924
97%
D-Sorbitol
95.0°C to 99.0°C
Authentic
C6H14O6
1533
+ 4.00 (20.00°C per eur. pharm.)
d-sorbitol, sorbitol, d-glucitol, glucitol, l-gulitol, --sorbitol, diakarmon, glucarine, sorbol, d---sorbitol
FBPFZTCFMRRESA-JGWLITMVSA-N
(2R,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol
5780
182.17
Granular Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-061-5

SDS
Documents

Documents

RUO – Research Use Only