missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Sorbitol, 97%

Catalog No. AC132730050
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
1 kg
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic bottle
Plastic drum
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC132730050 5 g Glass bottle
AC132731000 100 g Plastic bottle
AC132735000 500 g Plastic bottle
AC132730010 1 kg Plastic bottle
AC132730025 2.5 kg Plastic bottle
AC132730100 10 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. AC132730050 Supplier Thermo Scientific Chemicals Supplier No. 132730050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 50-70-4 | C6H14O6 | 182.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 50-70-4
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.17
MDL Number MFCD00004708
InChI Key FBPFZTCFMRRESA-JGWLITMVSA-N
Synonym d-sorbitol, sorbitol, d-glucitol, glucitol, l-gulitol, --sorbitol, diakarmon, glucarine, sorbol, d---sorbitol
PubChem CID 5780
ChEBI CHEBI:17924
IUPAC Name (2R,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol
SMILES C(C(C(C(C(CO)O)O)O)O)O

Specifications

Melting Point 95.0°C to 99.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (on anhydrous substance) (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 1533
Merck Index 158851
Specific Rotation Condition + 4.00 (20.00°C per eur. pharm.)
Specific Rotation + 4.00
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in acetic acid, soluble in glycerol and propylene glycol, slightly soluble in methanol, ethanol, phenol,, acetamide, acetone, dmf and pyridine, practically insoluble in most other org. sol
Formula Weight 182.17
Percent Purity 97%
Physical Form Granular Crystalline Powder
Chemical Name or Material D-Sorbitol
Show More Show Less
EINECSNumber 200-061-5

RUO – Research Use Only