missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals D-Sorbitol, 97%
CAS: 50-70-4 | C6H14O6 | 182.17 g/mol
Supplier: Thermo Scientific Chemicals 132730010
| Quantity | 1 kg |
|---|---|
| Packaging | Plastic bottle |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C6H14O6 | |
| MFCD00004708 | |
| d-sorbitol, sorbitol, d-glucitol, glucitol, l-gulitol, --sorbitol, diakarmon, glucarine, sorbol, d---sorbitol | |
| CHEBI:17924 | |
| C(C(C(C(C(CO)O)O)O)O)O |
Specifications
| D-Sorbitol | |
| 95.0°C to 99.0°C | |
| Authentic | |
| Plastic bottle | |
| 1 kg | |
| 1533 | |
| + 4.00 (20.00°C per eur. pharm.) | |
| d-sorbitol, sorbitol, d-glucitol, glucitol, l-gulitol, --sorbitol, diakarmon, glucarine, sorbol, d---sorbitol | |
| FBPFZTCFMRRESA-JGWLITMVSA-N | |
| (2R,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol | |
| 5780 | |
| 182.17 | |
| Granular Crystalline Powder |
| 50-70-4 | |
| White | |
| 96% min. (on anhydrous substance) (HPLC) | |
| C6H14O6 | |
| MFCD00004708 | |
| 158851 | |
| + 4.00 | |
| Solubility in water: soluble. Other solubilities: slightly soluble in acetic acid, soluble in glycerol and propylene glycol, slightly soluble in methanol, ethanol, phenol,, acetamide, acetone, dmf and pyridine, practically insoluble in most other org. sol | |
| C(C(C(C(C(CO)O)O)O)O)O | |
| 182.17 | |
| CHEBI:17924 | |
| 97% |
Safety and Handling
EINECSNumber : 200-061-5