missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Sorbitol, 97%

CAS: 50-70-4 | C6H14O6 | 182.17 g/mol

Supplier:  Thermo Scientific Chemicals 132731000

Quantity 100 g
Packaging Plastic bottle

 View more versions of this product

Catalog No. AC132731000


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

50-70-4
182.17
FBPFZTCFMRRESA-JGWLITMVSA-N
5780
(2R,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol
C6H14O6
MFCD00004708
d-sorbitol, sorbitol, d-glucitol, glucitol, l-gulitol, --sorbitol, diakarmon, glucarine, sorbol, d---sorbitol
CHEBI:17924
C(C(C(C(C(CO)O)O)O)O)O

Specifications

D-Sorbitol
95.0°C to 99.0°C
Authentic
Plastic bottle
100 g
1533
+ 4.00 (20.00°C per eur. pharm.)
d-sorbitol, sorbitol, d-glucitol, glucitol, l-gulitol, --sorbitol, diakarmon, glucarine, sorbol, d---sorbitol
FBPFZTCFMRRESA-JGWLITMVSA-N
(2R,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol
5780
182.17
Granular Crystalline Powder
50-70-4
White
96% min. (on anhydrous substance) (HPLC)
C6H14O6
MFCD00004708
158851
+ 4.00
Solubility in water: soluble. Other solubilities: slightly soluble in acetic acid, soluble in glycerol and propylene glycol, slightly soluble in methanol, ethanol, phenol,, acetamide, acetone, dmf and pyridine, practically insoluble in most other org. sol
C(C(C(C(C(CO)O)O)O)O)O
182.17
CHEBI:17924
97%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-061-5

SDS
Documents

Documents

Promotions

Promotions