missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals D(-)-Ribose, 99+%
CAS: 50-69-1 | C5H10O5 | 150.13 g/mol
$40.45 - $1,185.08
Chemical Identifiers
| CAS | 50-69-1 |
|---|---|
| Molecular Formula | C5H10O5 |
| Molecular Weight (g/mol) | 150.13 |
| MDL Number | MFCD00135453 |
| InChI Key | HMFHBZSHGGEWLO-ZHZWXSSZNA-N |
| Synonym | 2r,3r,4r-2,3,4,5-tetrahydroxypentanal, aldehydo-d-ribose, ribose, d, d--ribose, aldehydo-d-ribo-pentose, ribo-2,3,4,5-tetrahydroxyvaleraldehyde, d, d-ribo-2,3,4,5-tetrahydroxyvaleraldehyde, ribose 9ci, dsstox_cid_23917, dsstox_gsid_43917 |
| PubChem CID | 5311110 |
| ChEBI | CHEBI:47014 |
| SMILES | OC[C@H]1OC(O)[C@H](O)[C@@H]1O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC132360050
|
Thermo Scientific Chemicals
132360050 |
5 g | Glass bottle |
Each for $40.45
|
|
||||
|
AC132360250
|
Thermo Scientific Chemicals
132360250 |
25 g | Glass bottle |
Each for $97.20
|
|
||||
|
AC132361000
|
Thermo Scientific Chemicals
132361000 |
100 g | Glass bottle |
Each for $280.65
|
|
||||
|
AC132365000
|
Thermo Scientific Chemicals
132365000 |
500 g | Glass bottle |
Each for $1,185.08
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C5H10O5 | |
| MFCD00135453 | |
| 2r,3r,4r-2,3,4,5-tetrahydroxypentanal, aldehydo-d-ribose, ribose, d, d--ribose, aldehydo-d-ribo-pentose, ribo-2,3,4,5-tetrahydroxyvaleraldehyde, d, d-ribo-2,3,4,5-tetrahydroxyvaleraldehyde, ribose 9ci, dsstox_cid_23917, dsstox_gsid_43917 | |
| CHEBI:47014 |
Specifications
| 50-69-1 | |
| Beige to White or Yellow | |
| Authentic | |
| C5H10O5 | |
| 01, I, 434 | |
| − 20.00 (20.00°C c=4,H2O) | |
| 2r,3r,4r-2,3,4,5-tetrahydroxypentanal, aldehydo-d-ribose, ribose, d, d--ribose, aldehydo-d-ribo-pentose, ribo-2,3,4,5-tetrahydroxyvaleraldehyde, d, d-ribo-2,3,4,5-tetrahydroxyvaleraldehyde, ribose 9ci, dsstox_cid_23917, dsstox_gsid_43917 | |
| OC[C@H]1OC(O)[C@H](O)[C@@H]1O | |
| 150.13 | |
| CHEBI:47014 | |
| 99+% | |
| D(-)-Ribose |
| 88.0°C to 92.0°C | |
| 1% max. (40°C, 5 hrs, 20 torr) | |
| 99% min. (ELSD) (HPLC) | |
| MFCD00135453 | |
| 15, 8328 | |
| − 20.00 | |
| HMFHBZSHGGEWLO-ZHZWXSSZNA-N | |
| (2R,3R,4R)-2,3,4,5-tetrahydroxypentanal | |
| 5311110 | |
| 150.13 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : 200-059-4
RUO – Research Use Only