missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(-)-Ribose, 99+%

Catalog No. AC132365000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC132365000 500 g Glass bottle
AC132360050 5 g Glass bottle
AC132360250 25 g Glass bottle
AC132361000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC132365000 Supplier Thermo Scientific Chemicals Supplier No. 132365000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 50-69-1 | C5H10O5 | 150.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 50-69-1
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
MDL Number MFCD00135453
InChI Key HMFHBZSHGGEWLO-ZHZWXSSZNA-N
Synonym 2r,3r,4r-2,3,4,5-tetrahydroxypentanal, aldehydo-d-ribose, ribose, d, d--ribose, aldehydo-d-ribo-pentose, ribo-2,3,4,5-tetrahydroxyvaleraldehyde, d, d-ribo-2,3,4,5-tetrahydroxyvaleraldehyde, ribose 9ci, dsstox_cid_23917, dsstox_gsid_43917
PubChem CID 5311110
ChEBI CHEBI:47014
SMILES OC[C@H]1OC(O)[C@H](O)[C@@H]1O

Specifications

Melting Point 88.0°C to 92.0°C
Color Beige to White or Yellow
Loss on Drying 1% max. (40°C, 5 hrs, 20 torr)
Infrared Spectrum Authentic
Assay Percent Range 99% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 500 g
Beilstein 01, I, 434
Merck Index 15, 8328
Specific Rotation Condition − 20.00 (20.00°C c=4,H2O)
Specific Rotation − 20.00
IUPAC Name (2R,3R,4R)-2,3,4,5-tetrahydroxypentanal
Formula Weight 150.13
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material D(-)-Ribose
Show More Show Less
EINECSNumber 200-059-4

RUO – Research Use Only