accessibility menu, dialog, popup

UserName

(S)(R)(R)OOHOHHOHO

CAS RN 50-69-1

IUPAC Name: (3R,4S,5R)-5-(hydroxymethyl)oxolane-2,3,4-triol
Synonyms: D-ribofuranoside
Molecular Weight (g/mol): 150.13
Molecular Formula: C5H10O5
InChi Key: HMFHBZSHGGEWLO-ZHZWXSSZNA-N
SMILES: OC[C@H]1OC(O)[C@H](O)[C@@H]1O
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (3)
  • (4)
Physical Form 
  • (6)
  • (1)
Grade 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (1)
  • (2)
  • (7)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (4)
  • (6)
  • (1)
  • (1)
  • (2)