missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(-)-Ribose, 99+%

CAS: 50-69-1 | C5H10O5 | 150.13 g/mol

Supplier:  Thermo Scientific Chemicals 132361000

Quantity 100 g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC132361000


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

50-69-1
150.13
HMFHBZSHGGEWLO-ZHZWXSSZNA-N
5311110
OC[C@H]1OC(O)[C@H](O)[C@@H]1O
C5H10O5
MFCD00135453
2r,3r,4r-2,3,4,5-tetrahydroxypentanal, aldehydo-d-ribose, ribose, d, d--ribose, aldehydo-d-ribo-pentose, ribo-2,3,4,5-tetrahydroxyvaleraldehyde, d, d-ribo-2,3,4,5-tetrahydroxyvaleraldehyde, ribose 9ci, dsstox_cid_23917, dsstox_gsid_43917
CHEBI:47014

Specifications

D(-)-Ribose
88.0°C to 92.0°C
1% max. (40°C, 5 hrs, 20 torr)
99% min. (ELSD) (HPLC)
C5H10O5
MFCD00135453
15, 8328
− 20.00
HMFHBZSHGGEWLO-ZHZWXSSZNA-N
(2R,3R,4R)-2,3,4,5-tetrahydroxypentanal
5311110
150.13
Crystalline Powder
50-69-1
Beige to White or Yellow
Authentic
Glass bottle
100 g
01, I, 434
− 20.00 (20.00°C c=4,H2O)
2r,3r,4r-2,3,4,5-tetrahydroxypentanal, aldehydo-d-ribose, ribose, d, d--ribose, aldehydo-d-ribo-pentose, ribo-2,3,4,5-tetrahydroxyvaleraldehyde, d, d-ribo-2,3,4,5-tetrahydroxyvaleraldehyde, ribose 9ci, dsstox_cid_23917, dsstox_gsid_43917
OC[C@H]1OC(O)[C@H](O)[C@@H]1O
150.13
CHEBI:47014
99+%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-059-4

SDS
Documents

Documents

Promotions

Promotions