missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals D(-)-Ribose, 99+%
CAS: 50-69-1 | C5H10O5 | 150.13 g/mol
Supplier: Thermo Scientific Chemicals 132360250
| Quantity | 25 g |
|---|---|
| Packaging | Glass bottle |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C5H10O5 | |
| MFCD00135453 | |
| 2r,3r,4r-2,3,4,5-tetrahydroxypentanal, aldehydo-d-ribose, ribose, d, d--ribose, aldehydo-d-ribo-pentose, ribo-2,3,4,5-tetrahydroxyvaleraldehyde, d, d-ribo-2,3,4,5-tetrahydroxyvaleraldehyde, ribose 9ci, dsstox_cid_23917, dsstox_gsid_43917 | |
| CHEBI:47014 |
Specifications
| D(-)-Ribose | |
| 88.0°C to 92.0°C | |
| 1% max. (40°C, 5 hrs, 20 torr) | |
| 99% min. (ELSD) (HPLC) | |
| C5H10O5 | |
| MFCD00135453 | |
| 15, 8328 | |
| − 20.00 | |
| HMFHBZSHGGEWLO-ZHZWXSSZNA-N | |
| 5-(hydroxymethyl)oxolane-2,3,4-triol | |
| 5311110 | |
| 150.13 | |
| Crystalline Powder |
| 50-69-1 | |
| Beige to White or Yellow | |
| Authentic | |
| Glass bottle | |
| 25 g | |
| 01, I, 434 | |
| − 20.00 (20.00°C c=4,H2O) | |
| 2r,3r,4r-2,3,4,5-tetrahydroxypentanal, aldehydo-d-ribose, ribose, d, d--ribose, aldehydo-d-ribo-pentose, ribo-2,3,4,5-tetrahydroxyvaleraldehyde, d, d-ribo-2,3,4,5-tetrahydroxyvaleraldehyde, ribose 9ci, dsstox_cid_23917, dsstox_gsid_43917 | |
| OC[C@H]1OC(O)[C@H](O)[C@@H]1O | |
| 150.13 | |
| CHEBI:47014 | |
| 99+% |
Safety and Handling
EINECSNumber : 200-059-4